3-Sulfobenzoic acid monosodium salt 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium 3-sulphobenzoate, Sodium 3-sulfobenzoate

B23556-14 B23556-22 B23556-36
CAAAAB23556-14EA 42.5 CAD
CAAAAB23556-14 CAAAAB23556-22 CAAAAB23556-36
3-Sulfobenzoic acid monosodium salt 97%
3-Sulphobenzoic acid monosodium salt

Purity: 97% CAS No.: 17625-03-5 Molecular Formula: C7H5NaO5S Formula Weight: 224.17 MDL No.: MFCD00066378

Formula: C₇H₅NaO₅S
MW: 224.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066378
CAS Number: 17625-03-5
EINECS: 241-602-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR