4-(2-Methoxyethyl)phenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23548-09
B23548-18
CAAAAB23548-09EA
75.52
CAD
CAAAAB23548-09
CAAAAB23548-18
CAAAAB23548-30
4-(2-Methoxyethyl)phenol 98%
4-(2-Methoxyethyl)phenol
Purity: 98%
CAS No.: 56718-71-9
Molecular Formula: C9H12O2
Formula Weight: 152.19
Melting Point: 43-45°
MDL No.: MFCD00017537
Formula:
C₉H₁₂O₂ Melting Pt: 43…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017537 CAS Number: 56718-71-9 EINECS: 260-354-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...