2-(2-Aminothiazol-4-yl)acetic acid 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2-Amino-4-thiazolyl)acetic acid, 2-Amino-4-thiazoleacetic acid
B23534-14
B23534-22
CAAAAB23534-14EA
83.27
CAD
CAAAAB23534-14
CAAAAB23534-22
2-(2-Aminothiazol-4-yl)acetic acid 97%
2-(2-Aminothiazol-4-yl)acetic acid
Purity: 97%
CAS No.: 29676-71-9
Molecular Formula: C5H6N2O2S
Formula Weight: 158.18
Melting Point: ca 130° dec.
Beilstein Reference: 127415
MDL No.: MFCD00010068
Formula:
C₅H₆N₂O₂S MW: 158.18 g/mol Melting Pt: ca 130 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00010068 CAS Number: 29676-71-9 EINECS: 249-769-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...