3,5-Bis(trifluoromethyl)styrene 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23499-03
B23499-06
CAAAAB23499-03EA
162.74
CAD
CAAAAB23499-03
CAAAAB23499-06
3,5-Bis(trifluoromethyl)styrene 98% stabilized
3,5-Bis(trifluoromethyl)styrene
Purity: 98%
CAS No.: 349-59-7
Molecular Formula: C10H6F6
Formula Weight: 240.15
Storage Temperature: 2°C to 8°C
Flash Point: 50°(122°F)
Refractive Index: 1.4250
Melting Point: 7-8°
Boiling Point: 59-61°/20mm
UN No.: 1993
MDL No.: MFCD00075538
Formula:
C₁₀H₆F₆ Boiling Pt: 59…61 °C/20 mm Melting Pt: 7…8 °C Density: 1.334 Flash Pt: 50 °C(122 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00075538 CAS Number: 349-59-7 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...