Benzyl methyl ether 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23490-09
B23490-18
CAAAAB23490-09EA
63.67
CAD
CAAAAB23490-09
CAAAAB23490-18
CAAAAB23490-30
Benzyl methyl ether 97%
Benzyl methyl ether
Purity: 99%
CAS No.: 538-86-3
Molecular Formula: C6H5CH2OCH3
Formula Weight: 122.17
Flash Point: 56°(133°F)
Refractive Index: 1.5015
Melting Point: -53°
Boiling Point: 173-175°
Merck Reference: 14,1140
UN No.: 3271
MDL No.: MFCD00025901
Formula:
C₈H₁₀O Boiling Pt: 173…175 °C Melting Pt: –53 °C Density: 0.940 Flash Pt: 56 °C(133 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00025901 CAS Number: 538-86-3 EINECS: 208-705-7 UN: 3271 ADR: "3",III Merck Index: 14,1140 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...