S-(Carboxymethyl)-L-cysteine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23487-14
B23487-22
CAAAB23487-14EA
70.94
CAD
CAAAB23487-14
CAAAB23487-22
S-(Carboxymethyl)-L-cysteine 97%
S-(Carboxymethyl)-L-cysteine
Purity: 97%
CAS No.: 638-23-3
Molecular Formula: C5H9NO4S
Formula Weight: 179.20
Storage Temperature: 2°C to 8°C
Melting Point: ca 200° dec.
Beilstein Reference: 1725012
Optical Rotation: -29° (c=10 in 10% NaHCO)
MDL No.: MFCD00002614
Formula:
C₅H₉NO₄S Melting Pt: ca 200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002614 CAS Number: 638-23-3 EINECS: 211-327-5 Merck Index: 14,1802 |
Specification Test Results
Optical rotation | -29° (c=10 in 10% NaHCOƒ) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...