4-Chloro-3-nitrobenzophenone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23451-14
B23451-22
CAAAB23451-14EA
47.87
CAD
CAAAB23451-14
CAAAB23451-22
4-Chloro-3-nitrobenzophenone 99%
4-Chloro-3-nitrobenzophenone
Purity: 99%
CAS No.: 56107-02-9
Molecular Formula: C13H8ClNO3
Formula Weight: 261.66
Melting Point: 104-106°
Boiling Point: 233-235°/12mm
MDL No.: MFCD00007081
Formula:
C₁₃H₈ClNO₃ Boiling Pt: 233…235 °C/12 mm Melting Pt: 104…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007081 CAS Number: 56107-02-9 EINECS: 259-995-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...