2-Sulfobenzoic acid hydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-sulphobenzoic acid hydrate, 2-sulfobenzoic acid hydrate

B23381-14 B23381-22
CAAAB23381-14EA 66.36 CAD
CAAAB23381-14 CAAAB23381-22
2-Sulfobenzoic acid hydrate 98%
2-Sulphobenzoic acid hydrate

Purity: 98% CAS No.: 123333-68-6 Molecular Formula: C7H6O5S.xH2O Formula Weight: 202.19(anhy) Melting Point: ca 68-72° Beilstein Reference: 2106852 UN No.: 3261 MDL No.: MFCD00149534

Formula: C₇H₆O₅S·H₂O
MW: 202.19 g/mol (anhy)
Melting Pt: ∼ 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00149534
CAS Number: 123333-68-6
EINECS: 211-176-5
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR