1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt 90+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Hydroxy-4-nitroso-2,7-naphthalenedisulphonic acid disodium salt, Nitroso-R salt

B23375-14 B23375-22 B23375-36
CAAAAB23375-14EA 75.37 CAD
CAAAAB23375-14 CAAAAB23375-22 CAAAAB23375-36
1-Nitroso-2-naphthol-3,6-disulfonic acid disodium salt 90+%
1-Nitroso-2-naphthol-3,6-disulphonic acid disodium salt

Purity: 90+% CAS No.: 525-05-3 Molecular Formula: C10H5NNa2O8S2 Formula Weight: 377.26 Melting Point: 300° Beilstein Reference: 3643930 Merck Reference: 14,6644 MDL No.: MFCD00003885

Formula: C₁₀H₅NNa₂O₈S₂
MW: 377.26 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003885
CAS Number: 525-05-3
EINECS: 208-369-1
Merck Index: 14,6644

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR