3-Nitrobenzylamine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

B23349-03 B23349-06 B23349-14
CAAAAB23349-03EA 86.9 CAD
CAAAAB23349-03 CAAAAB23349-06 CAAAAB23349-14
3-Nitrobenzylamine hydrochloride 97%
3-Nitrobenzylamine hydrochloride

Purity: 97% CAS No.: 26177-43-5 Molecular Formula: C7H8N2O2.HCl Formula Weight: 188.62 Melting Point: ca 225° dec. MDL No.: MFCD00012858

Formula: C₇H₉ClN₂O₂
Melting Pt: ca 225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012858
CAS Number: 26177-43-5
EINECS: 247-502-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR