(3-Nitrophenyl)methanesulfonyl chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Nitro-α-toluenesulphonyl chloride, 3-Nitro-α-toluenesulfonyl chloride

B23333-03 B23333-06 B23333-14
CAAAB23333-03EA 106.49 CAD
CAAAB23333-03 CAAAB23333-06 CAAAB23333-14
(3-Nitrophenyl)methanesulfonyl chloride 95%
(3-Nitrophenyl)methanesulphonyl chloride

Purity: 95% CAS No.: 58032-84-1 Molecular Formula: C7H6ClNO4S Formula Weight: 235.64 Melting Point: 95-102° UN No.: 3261 MDL No.: MFCD01859935

Formula: C₇H₆ClNO₄S
MW: 235.64 g/mol
Melting Pt: 95…102 °C
Storage Temperature: Ambient
MDL Number: MFCD01859935
CAS Number: 58032-84-1
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR