Zirconium(IV) butoxide ≥80% (w/w) in 1-butanol
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23317-0B
B23317-18
B23317-30
CAAAAB23317-0BEA
262.28
CAD
CAAAAB23317-0B
CAAAAB23317-18
CAAAAB23317-30
Zirconium(IV) butoxide ≥80% (w/w) in 1-butanol
Zirconium(IV) butoxide
CAS No.: 1071-76-7
Molecular Formula: C16H36O4Zr
Formula Weight: 383.68
Flash Point: 38°(100°F)
Refractive Index: 1.466
Odor: Alcohol-like
Beilstein Reference: 4132649
UN No.: 1120
MDL No.: MFCD00048765
Formula:
C₁₆H₃₆O₄Zr Density: 1.070 Flash Pt: 38 °C(100 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00048765 CAS Number: 1071-76-7 EINECS: 213-995-3 UN: 1120 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...