2-Chloroethyl phenyl sulfide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23265-06
B23265-14
CAAAB23265-06EA
58.93
CAD
CAAAB23265-06
CAAAB23265-14
2-Chloroethyl phenyl sulfide 98%
2-Chloroethyl phenyl sulphide
Purity: 98%
CAS No.: 5535-49-9
Molecular Formula: C8H9ClS
Formula Weight: 172.68
Flash Point: 104°(220°F)
Refractive Index: 1.5830
Boiling Point: 245°
Beilstein Reference: 1907116
UN No.: 2922
MDL No.: MFCD00013691
Formula:
C₈H₉ClS Boiling Pt: 245 °C Density: 1.178 Flash Pt: 104 °C(220 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013691 CAS Number: 5535-49-9 EINECS: 226-891-8 UN: 2922 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...