Triethyl-2-phosphonopropionate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23261-09
B23261-18
CAAAAB23261-09EA
79.16
CAD
CAAAAB23261-09
CAAAAB23261-18
Triethyl-2-phosphonopropionate 98%
Triethyl 2-phosphonopropionate
Purity: 98%
CAS No.: 3699-66-9
Molecular Formula: C9H19O5P
Formula Weight: 238.22
Flash Point: 88°(190°F)
Refractive Index: 1.4320
Odor: Odorless
Boiling Point: 143-144°/12mm
Beilstein Reference: 1786994
MDL No.: MFCD00009159
Formula:
C₉H₁₉O₅P Boiling Pt: 143…144 °C (12 mmHg) Density: 1.111 Flash Pt: 88 °C (190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009159 CAS Number: 3699-66-9 EINECS: 223-033-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...