Bathophenanthrolinedisulfonic acid disodium salt hydrate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4,7-Diphenyl-1,10-phenanthrolinedisulphonic acid disodium salt hydrate, 4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid disodium salt hydrate

B23244-03 B23244-06 B23244-MD
CAAAAB23244-03EA 188.02 CAD
CAAAAB23244-03 CAAAAB23244-06 CAAAAB23244-MD
Bathophenanthrolinedisulfonic acid disodium salt hydrate 98%
Bathophenanthrolinedisulphonic acid disodium salt hydrate

Purity: 98% CAS No.: 52746-49-3 Molecular Formula: C24H14N2Na2O6S2.xH2O Formula Weight: 536.48(anhy) Melting Point: 300° MDL No.: MFCD00149310

Formula: C₂₄H₁₄N₂Na₂O₆S₂·H₂O
MW: 536.48 g/mol (anhy)
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149310
CAS Number: 52746-49-3
EINECS: 258-152-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR