Methyl-2-(chlorosulfonyl)benzoate 94%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23211-06
B23211-14
CAAAB23211-06EA
113.76
CAD
CAAAB23211-06
CAAAB23211-14
CAAAB23211-22
Methyl-2-(chlorosulfonyl)benzoate 94%
Methyl 2-(chlorosulphonyl)benzoate
Purity: 94%
CAS No.: 26638-43-7
Molecular Formula: C8H7ClO4S
Formula Weight: 234.66
Storage Temperature: 2°C to 8°C
Melting Point: 59-64°
UN No.: 3261
MDL No.: MFCD00009797
Formula:
C₈H₇ClO₄S Melting Pt: 59…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009797 CAS Number: 26638-43-7 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...