Ethyl dibromoacetate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23209-06
B23209-14
CAAAB23209-06EA
167.48
CAD
CAAAB23209-06
CAAAB23209-14
Ethyl dibromoacetate 96%
Ethyl dibromoacetate
Purity: 96%
CAS No.: 617-33-4
Molecular Formula: C4H6Br2O2
Formula Weight: 245.90
Refractive Index: 1.4990
Boiling Point: 76-78°/12mm
MDL No.: MFCD00041718
Formula:
C₄H₆Br₂O₂ Boiling Pt: 76…78 °C/12 mm Density: 1.902 Storage Temperature: Ambient |
MDL Number:
MFCD00041718 CAS Number: 617-33-4 EINECS: 210-510-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...