3-(2-Aminoethylamino)propylmethyldimethoxysilane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-(2-Aminoethylamino)propyldimethoxymethylsilane
B23159-14
B23159-22
B23159-36
CAAAAB23159-14EA
54.19
CAD
CAAAAB23159-14
CAAAAB23159-22
CAAAAB23159-36
3-(2-Aminoethylamino)propylmethyldimethoxysilane 97%
3-(2-Aminoethylamino)propylmethyldimethoxysilane
Purity: 97%
CAS No.: 3069-29-2
Molecular Formula: (CH3O)2(CH3)Si(CH2)3NHCH2CH2NH2
Formula Weight: 206.36
Flash Point: 90°(194°F)
Refractive Index: 1.450
Odor: Amine-like
Boiling Point: 265°
Beilstein Reference: 2074259
UN No.: 2735
MDL No.: MFCD00054776
Formula:
C₈H₂₂N₂O₂Si Boiling Pt: 265 °C Density: 0.968 Flash Pt: 90 °C(194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00054776 CAS Number: 3069-29-2 EINECS: 221-336-6 UN: 2735 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...