(1R,3S)-(+)-Camphoric acid 98+%
Supplier: Thermo Scientific Chemicals
B23106-14
B23106-22
B23106-36
CAAAB23106-14EA
51.35
CAD
CAAAB23106-14
CAAAB23106-22
CAAAB23106-36
(1R,3S)-(+)-Camphoric acid 98+%
(1R,3S)-(+)-Camphoric acid
Purity: 98+%
CAS No.: 124-83-4
Molecular Formula: C10H16O4
Formula Weight: 200.23
Melting Point: 183-188°
Beilstein Reference: 2050204
Merck Reference: 14,1733
Optical Rotation: +46° (c=1 in methanol)
MDL No.: MFCD00001375
Formula:
C₁₀H₁₆O₄ Melting Pt: 183…188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001375 CAS Number: 124-83-4 EINECS: 204-715-0 Merck Index: 14,1733 |
Specification Test Results
Optical rotation | +46° (c=1 in methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...