Benzyl ethyl malonate ≥85%, tech.
Supplier: Thermo Scientific Chemicals
B23011-14
B23011-22
CAAAB23011-06EA
474
CAD
CAAAB23011-06
CAAAB23011-14
CAAAB23011-22
Benzyl ethyl malonate ≥85%, tech.
Benzyl ethyl malonate
Purity: 85%
CAS No.: 42998-51-6
Molecular Formula: C12H14O4
Formula Weight: 222.24
Refractive Index: 1.4970
Boiling Point: 145°/4mm
MDL No.: MFCD00009194
Formula:
C₁₂H₁₄O₄ Boiling Pt: 145 °C/4 mm Storage Temperature: Ambient |
MDL Number:
MFCD00009194 CAS Number: 42998-51-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...