Ethyl-3-phenylglycidate (mixture of cis and trans isomers) 90+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22990-AD
B22990-AK
CAAAB22990-ADEA
60.67
CAD
CAAAB22990-AD
CAAAB22990-AK
CAAAB22990-AU
Ethyl-3-phenylglycidate (mixture of cis and trans isomers) 90+%
Ethyl 3-phenylglycidate (mixture of cis and trans isomers)
Purity: 90+%
CAS No.: 121-39-1
Molecular Formula: C11H12O3
Formula Weight: 192.21
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₁₁H₁₂O₃ Boiling Pt: 95…97 °C/0.5 mm Density: 1.102 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005123 CAS Number: 121-39-1 EINECS: 204-467-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...