3-(Chlorosulfonyl)benzoic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

B22987-06 B22987-14
CAAAB22987-06EA 52.77 CAD
CAAAB22987-06 CAAAB22987-14 CAAAB22987-22
3-(Chlorosulfonyl)benzoic acid 97%
3-(Chlorosulphonyl)benzoic acid

Purity: 97% CAS No.: 4025-64-3 Molecular Formula: C7H5ClO4S Formula Weight: 220.63 Melting Point: 133-138° Beilstein Reference: 2645465 UN No.: 3261 MDL No.: MFCD00024877

Formula: C₇H₅ClO₄S
MW: 220.63 g/mol
Melting Pt: 133…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00024877
CAS Number: 4025-64-3
EINECS: 223-692-8
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR