L(-)-Bornyl acetate 95%
Supplier: Thermo Scientific Chemicals
Warning
|
B22957-14
B22957-22
B22957-36
CAAAB22957-14EA
38.55
CAD
CAAAB22957-14
CAAAB22957-22
CAAAB22957-36
L(-)-Bornyl acetate 95%
L(-)-Bornyl acetate
Purity: 95%
CAS No.: 5655-61-8
Molecular Formula: C12H20O2
Formula Weight: 196.29
Flash Point: 84°(184°F)
Refractive Index: 1.4630
Boiling Point: 223-224°
Beilstein Reference: 2502036
Merck Reference: 14,1339
MDL No.: MFCD00135942
Formula:
C₁₂H₂₀O₂ Boiling Pt: 223…224 °C Density: 0.980 Flash Pt: 84 °C(184 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00867808 CAS Number: 5655-61-8 EINECS: 227-101-4 Merck Index: 14,1339 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...