2-(Methylthio)ethylamine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22941-03
B22941-06
CAAAB22941-03EA
116.45
CAD
CAAAB22941-03
CAAAB22941-06
2-(Methylthio)ethylamine 95%
2-(Methylthio)ethylamine
Purity: 95%
CAS No.: 18542-42-2
Molecular Formula: C3H9NS
Formula Weight: 91.18
Flash Point: 36°(96°F)
Refractive Index: 1.4965
Odor: Amine-like
Boiling Point: 146-148°
Beilstein Reference: 1731099
UN No.: 2734
MDL No.: MFCD00014825
Formula:
C₃H₉NS Boiling Pt: 146…148 °C Density: 0.974 Flash Pt: 36 °C (96 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014825 CAS Number: 18542-42-2 EINECS: 242-412-5 UN: 2734 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...