Ammonium cobalt(II) phosphate, anhydrous 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22934-18
B22934-30
CAAAAB22934-18EA
105.39
CAD
CAAAAB22934-18
CAAAAB22934-30
Ammonium cobalt(II) phosphate, anhydrous 98%
Ammonium cobalt(II) phosphate
Purity: 98%
CAS No.: 14590-13-7
Molecular Formula: H4CoNO4P
Formula Weight: 171.94
MDL No.: MFCD02683100
Formula:
H₄CoNO₄P MW: 171.94 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02683100 CAS Number: 14590-13-7 EINECS: 238-633-1 Merck Index: 14,513 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...