2,4,6-Triisopropylphenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,4,6-Triisopropylbenzeneboronic acid, (2,4,6-Triisopropylphenyl)boronic acid

B22891-03 B22891-06 B22891-14
CAAAB22891-03EA 113.13 CAD
CAAAB22891-03 CAAAB22891-06 CAAAB22891-14
2,4,6-Triisopropylphenylboronic acid 98%
2,4,6-Triisopropylphenylboronic acid

Purity: 98% CAS No.: 154549-38-9 Molecular Formula: C15H25BO2 Formula Weight: 248.16 Melting Point: ca 165° dec. MDL No.: MFCD02683099

Formula: C₁₅H₂₅BO₂
MW: 248.16 g/mol
Melting Pt: ca 165 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02683099
CAS Number: 154549-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR