N-[Tris(hydroxymethyl)methyl]acrylamide 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(1,3-Dihydroxy-2-(hydroxymethyl)propan-2-yl)acrylamide

B22854-06 B22854-14 B22854-22
CAAAB22854-06EA 154.05 CAD
CAAAB22854-06 CAAAB22854-14 CAAAB22854-22
N-[Tris(hydroxymethyl)methyl]acrylamide 99%
N-[Tris(hydroxymethyl)methyl]acrylamide

Purity: 99% CAS No.: 13880-05-2 Molecular Formula: C7H13NO4 Formula Weight: 175.18 Storage Temperature: 2°C to 8°C Melting Point: 134-138° MDL No.: MFCD00081045

Formula: C₇H₁₃NO₄
MW: 175.18 g/mol
Melting Pt: 134…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00081045
CAS Number: 13880-05-2
EINECS: 237-645-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR