Pyridino[3,2-b]pyrazine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22843-02
B22843-88
CAAAB22843-02EA
117.08
CAD
CAAAB22843-02
CAAAB22843-88
Pyridino[3,2-b]pyrazine 98%
Pyridino[3,2-b]pyrazine
Purity: 98%
CAS No.: 322-46-3
Molecular Formula: C7H5N3
Formula Weight: 131.14
Melting Point: 142-144°
MDL No.: MFCD00006717
Formula:
C₇H₅N₃ Melting Pt: 142…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006717 CAS Number: 322-46-3 EINECS: 206-294-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...