Bathocuproin 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline

B22841-03 B22841-06
CAAAAB22841-03EA 146.62 CAD
CAAAAB22841-03 CAAAAB22841-06
Bathocuproin 98%
Bathocuproin

Purity: 98% CAS No.: 4733-39-5 Molecular Formula: C26H20N2 Formula Weight: 360.46 Melting Point: 279-285° Beilstein Reference: 306714

Formula: C₂₆H₂₀N₂
MW: 360.46 g/mol
Melting Pt: 279…285 °C
Storage Temperature: Ambient
MDL Number: MFCD00004972
CAS Number: 4733-39-5
EINECS: 225-240-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR