5-Chlorothiophene-2-carboxaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-5-thiophenecarboxaldehyde, 2-Chloro-5-formylthiophene

B22833-03 B22833-06
CAAAB22833-03EA 82.95 CAD
CAAAB22833-03 CAAAB22833-06 CAAAB22833-14
5-Chlorothiophene-2-carboxaldehyde 97%
5-Chlorothiophene-2-carboxaldehyde

Purity: 97% CAS No.: 7283-96-7 Molecular Formula: C5H3ClOS Formula Weight: 46.60 Storage Temperature: 2°C to 8°C Flash Point: 97°(208°F) Refractive Index: 1.6040 Boiling Point: 88-90°/9mm MDL No.: MFCD00047090

Formula: C₅H₃ClOS
MW: 146.59 g/mol
Boiling Pt: 88…90 °C/9 mm
Density: 1.376
Flash Pt: 97 °C(208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00047090
CAS Number: 7283-96-7
EINECS: 230-708-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR