4-Methoxy-4'-nitrobenzophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Methoxyphenyl)(4-nitrophenyl)methanone

B22720-03 B22720-06 B22720-14
CAAAB22720-03EA 85.16 CAD
CAAAB22720-03 CAAAB22720-06 CAAAB22720-14
4-Methoxy-4'-nitrobenzophenone 97%
4-Methoxy-4'-nitrobenzophenone

Purity: 97% CAS No.: 1151-94-6 Molecular Formula: C14H11NO4 Formula Weight: 257.25 Melting Point: 125-127° MDL No.: MFCD00178348

Formula: C₁₄H₁₁NO₄
MW: 257.25 g/mol
Melting Pt: 125…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00178348
CAS Number: 1151-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR