Boc-D-Alanine 98
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Boc-D-alanine, Boc-D-Ala-OH
B22706-06
B22706-14
CAAAB22706-06EA
67.62
CAD
CAAAB22706-06
CAAAB22706-14
Boc-D-Alanine 98
Boc-D-Alanine
Purity: 98+%
CAS No.: 7764-95-6
Molecular Formula: C8H15NO4
Formula Weight: 189.21
Melting Point: 81-84°
Beilstein Reference: 2048396
Optical Rotation: +26° (c=2 in acetic acid)
MDL No.: MFCD00063123
Formula:
C8H15NO4 MW: 189.2 g/mol Melting Pt: 81…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063123 CAS Number: 7764-95-6 |
Specification Test Results
Optical rotation | +26° (c=2 in acetic acid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...