Methyl-3-methoxy-4-methylbenzoate 98%

Supplier: Thermo Scientific Chemicals

B22695-06 B22695-14
CAAAB22695-03EA 70.45 CAD
CAAAB22695-03 CAAAB22695-06 CAAAB22695-14
Methyl-3-methoxy-4-methylbenzoate 98%
Methyl 3-methoxy-4-methylbenzoate

Purity: 98% CAS No.: 3556-83-0 Molecular Formula: C10H12O3 Formula Weight: 180.21 Flash Point: 110°(230°F) Refractive Index: 1.5295 Melting Point: 47-50° Boiling Point: 119-120°/1mm Beilstein Reference: 1948339 MDL No.: MFCD00082710

Formula: C₁₀H₁₂O₃
Boiling Pt: 119…120 °C/1 mm
Melting Pt: 47…50 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00082710
CAS Number: 3556-83-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR