L-Tyrosine methyl ester hydrochloride ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22683-14
B22683-22
CAAAB22683-14EA
90.85
CAD
CAAAB22683-14
CAAAB22683-22
L-Tyrosine methyl ester hydrochloride ≥98%
L-Tyrosine methyl ester hydrochloride
Purity: 98+%
CAS No.: 3417-91-2
Molecular Formula: C10H13NO3.HCl
Formula Weight: 231.68
Storage Temperature: 2°C to 8°C
Melting Point: 187° dec.
Beilstein Reference: 3917353
Optical Rotation: -5.2° (c= 2.4 in water)
MDL No.: MFCD00012607
Formula:
C₁₀H₁₃NO₃ Melting Pt: 187 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012607 CAS Number: 3417-91-2 EINECS: 222-313-3 |
Specification Test Results
Optical rotation | -5.2° (c= 2.4 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...