4-(Methylthio)butanol 99%
Supplier: Thermo Scientific Chemicals
B22673-03
B22673-06
B22673-14
CAAAB22673-03EA
60.51
CAD
CAAAB22673-03
CAAAB22673-06
CAAAB22673-14
4-(Methylthio)butanol 99%
4-(Methylthio)butanol
Purity: 99%
CAS No.: 20582-85-8
Molecular Formula: C5H12OS
Formula Weight: 120.21
Flash Point: 101°(215°F)
Refractive Index: 1.4870
Odor: Unpleasant
Boiling Point: 95-98°/20mm
MDL No.: MFCD00002973
Formula:
C₅H₁₂OS Boiling Pt: 95…98 °C/20 mm Density: 0.999 Flash Pt: 101 °C(215 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002973 CAS Number: 20582-85-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...