N-Carbobenzoxy-L-serine benzyl ester 99%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Cbz-L-serine Benzyl Ester, Z-Ser-OBzl, Benzyl ((benzyloxy)carbonyl)-L-serinate

B22647-03 B22647-06
CAAAB22647-03EA 80.42 CAD
CAAAB22647-03 CAAAB22647-06
N-Carbobenzoxy-L-serine benzyl ester 99%
N-Carbobenzoxy-L-serine benzyl ester

Purity: 99% CAS No.: 21209-51-8 Molecular Formula: C18H19NO5 Formula Weight: 329.32 Melting Point: 82-84° MDL No.: MFCD00037824

Formula: C₁₈H₁₉NO₅
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00037824
CAS Number: 21209-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR