Hydroquinone-O,O'-diacetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B22645-06 B22645-14 B22645-22
CAAAB22645-06EA 80.26 CAD
CAAAB22645-06 CAAAB22645-14 CAAAB22645-22
Hydroquinone-O,O'-diacetic acid 98%
Hydroquinone-O,O'-diacetic acid

Purity: 98% CAS No.: 2245-53-6 Molecular Formula: C10H10O6 Formula Weight: 226.19 Melting Point: ca 255° dec. Beilstein Reference: 1984454 MDL No.: MFCD00016816

Formula: C₁₀H₁₀O₆
MW: 226.19 g/mol
Melting Pt: ∼ 255 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00016816
CAS Number: 2245-53-6
EINECS: 218-834-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR