2-Mercaptothiophene 97% (as monomer) contains dimer

Supplier: Thermo Scientific Chemicals
Warning

B22642-03 B22642-06 B22642-14
CAAAAB22642-03EA 62.73 CAD
CAAAAB22642-03 CAAAB22642-06 CAAAB22642-14
2-Mercaptothiophene 97% (as monomer) contains dimer
2-Mercaptothiophene

Purity: 97% CAS No.: 7774-74-5 Molecular Formula: C4H4S2 Formula Weight: 116.20 Storage Temperature: 2°C to 8°C Refractive Index: 1.6170 Odor: Unpleasant Boiling Point: 62-65°/12mm Beilstein Reference: 104650 MDL No.: MFCD00051666

Formula: C₄H₄S₂
Boiling Pt: 62…65 °C/12 mm
Density: 1.247
Flash Pt: 66 °C(151 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00051666
CAS Number: 7774-74-5
EINECS: 231-881-1
UN: 3334
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR