7-Oxabicyclo[2.2.1]heptane 98%

Supplier: Thermo Scientific Chemicals
Danger

B22612-06 B22612-14 B22612-22
CAAAB22612-06EA 172.22 CAD
CAAAB22612-06 CAAAB22612-14 CAAAB22612-22
7-Oxabicyclo[2.2.1]heptane 98%
7-Oxabicyclo[2.2.1]heptane

Purity: 98% CAS No.: 279-49-2 Molecular Formula: C6H10O Formula Weight: 98.15 Flash Point: 12°(55°F) Refractive Index: 1.4480 Boiling Point: 118-120°/713mm UN No.: 3271 MDL No.: MFCD00082342

Formula: C₆H₁₀O
MW: 98.15 g/mol
Boiling Pt: 118…120 °C/713 mm
Density: 0.968
Flash Pt: 12 °C(55 °F)
Storage Temperature: Ambient
MDL Number: MFCD00082342
CAS Number: 279-49-2
EINECS: 205-998-3
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR