4-Cyano-4''-n-pentyl-p-terphenyl 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4''-Pentyl-[1,1':4',1''-terphenyl]-4-carbonitrile, 4-Cyano-4''-pentyl-p-terphenyl

B22607-02 B22607-88
CAAAB22607-02EA 109.97 CAD
CAAAB22607-02 CAAAB22607-88
4-Cyano-4''-n-pentyl-p-terphenyl 99%
4-Cyano-4''-n-pentyl-p-terphenyl

Purity: 99% CAS No.: 54211-46-0 Molecular Formula: C24H23N Formula Weight: 325.45 Melting Point: 128-130° UN No.: 3439 MDL No.: MFCD00799423

Formula: C₂₄H₂₃N
MW: 325.45 g/mol
Melting Pt: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00799423
CAS Number: 54211-46-0
EINECS: 259-028-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR