2-Fluoro-α-methyl-4-biphenylacetic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

B22603-03 B22603-06 B22603-14
CAAAB22603-03EA 62.57 CAD
CAAAB22603-03 CAAAB22603-06 CAAAB22603-14
2-Fluoro-α-methyl-4-biphenylacetic acid 99%
2-Fluoro-α-methyl-4-biphenylacetic acid

A cyclooxygenase inhibitor


Purity: 99% CAS No.: 5104-49-4 Molecular Formula: C15H13FO2 Formula Weight: 244.27 Odor: Odorless Melting Point: 112-114° UN No.: 2811 MDL No.: MFCD00079303

Formula: C₁₅H₁₃FO₂
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00079303
CAS Number: 5104-49-4
EINECS: 225-827-6
UN: 2811
ADR: 6.1,III
Merck Index: 14,4199

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR