2-Methylbutyryl chloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B22599-06
B22599-14
B22599-22
CAAAB22599-06EA
93.69
CAD
CAAAB22599-06
CAAAB22599-14
CAAAB22599-22
2-Methylbutyryl chloride 99%
2-Methylbutyryl chloride
Purity: 99%
CAS No.: 57526-28-0
Molecular Formula: C5H9ClO
Formula Weight: 120.58
Flash Point: 20°(68°F)
Refractive Index: 1.4180
Boiling Point: 116-117°
UN No.: 2924
MDL No.: MFCD00041726
Formula:
C₅H₉ClO Boiling Pt: 116…117 °C Density: 0.98 Flash Pt: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041726 CAS Number: 57526-28-0 EINECS: 260-787-3 UN: 2924 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...