5-Methoxy-2-benzimidazolinone 98%
Supplier: Thermo Scientific Chemicals
B22580-03
B22580-06
CAAAB22580-03EA
87.22
CAD
CAAAB22580-03
CAAAB22580-06
5-Methoxy-2-benzimidazolinone 98%
5-Methoxy-2-benzimidazolinone
Purity: 98%
CAS No.: 2080-75-3
Molecular Formula: C8H8N2O2
Formula Weight: 164.17
Melting Point: ca 255° dec.
Beilstein Reference: 141043
MDL No.: MFCD00051946
Formula:
C₈H₈N₂O₂ Melting Pt: ca 255 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00051946 CAS Number: 2080-75-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...