2-Mercapto-1,3,4-thiadiazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3,4-Thiadiazole-2-thiol

B22556-09 B22556-18
CAAAB22556-09EA 79.95 CAD
CAAAB22556-09 CAAAB22556-18 CAAAB22556-30
2-Mercapto-1,3,4-thiadiazole 98%
2-Mercapto-1,3,4-thiadiazole

Purity: 98% CAS No.: 18686-82-3 Molecular Formula: C2H2N2S2 Formula Weight: 118.17 Melting Point: 142-144° MDL No.: MFCD00040404

Formula: C₂H₂N₂S₂
MW: 118.17 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00040404
CAS Number: 18686-82-3
EINECS: 242-504-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR