4-(4-Chlorobenzoyl)pyridine 99+%
Supplier: Thermo Scientific Chemicals
B22551-03
B22551-06
CAAAB22551-03EA
100.96
CAD
CAAAB22551-03
CAAAB22551-06
CAAAB22551-14
4-(4-Chlorobenzoyl)pyridine 99+%
4-(4-Chlorobenzoyl)pyridine
Purity: 99+%
CAS No.: 14548-48-2
Molecular Formula: C12H8ClNO
Formula Weight: 217.66
Melting Point: 107-109°
MDL No.: MFCD00006431
Formula:
C₁₂H₈ClNO Melting Pt: 107…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006431 CAS Number: 14548-48-2 EINECS: 238-587-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...