Bathocuproin sulfonate disodium salt hydrate 97%

Supplier: Thermo Scientific Chemicals

B22550-03 B22550-06 B22550-MD
CAAAAB22550-03EA 189.6 CAD
CAAAAB22550-03 CAAAAB22550-06 CAAAAB22550-MD
Bathocuproin sulfonate disodium salt hydrate 97%
Bathocuproin sulphonate disodium salt hydrate

Used as a stain for proteins in polyacrylamide gels; for determination of iron and copper; and uric acid by automated procedures


Purity: 97% CAS No.: 52698-84-7 Molecular Formula: C26H18N2Na2O6S2.xH2O Formula Weight: 564.55(anhy) Melting Point: 300° Beilstein Reference: 9033637

Formula: C₂₆H₁₈N₂Na₂O₆S₂·H₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD03012947
CAS Number: 52698-84-7
EINECS: 258-111-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR