Ethyl-3-amino-4,4,4-trifluorocrotonate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B22545-06
B22545-14
CAAAB22545-06EA
56.72
CAD
CAAAB22545-06
CAAAB22545-14
Ethyl-3-amino-4,4,4-trifluorocrotonate 97%
Ethyl 3-amino-4,4,4-trifluorocrotonate
Purity: 97%
CAS No.: 372-29-2
Molecular Formula: C6H8F3NO2
Formula Weight: 183.13
Flash Point: 65°(149°F)
Refractive Index: 1.4229
Odor: Amine-like
Melting Point: 24-26°
Boiling Point: 83-85°/15mm
Beilstein Reference: 4397839
UN No.: 3259
MDL No.: MFCD00068195
Formula:
C₆H₈F₃NO₂ Boiling Pt: 83…85 °C/15 mm Melting Pt: 24…26 °C Density: 1.245 Flash Pt: 65 °C(149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00068195 CAS Number: 372-29-2 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...