Benzyl phenyl ether 97%

Supplier: Thermo Scientific Chemicals
Warning

B22539-06 B22539-14 B22539-22
CAAAB22539-06EA 48.66 CAD
CAAAB22539-06 CAAAB22539-14 CAAAB22539-22
Benzyl phenyl ether 97%
Benzyl phenyl ether

Purity: 97% CAS No.: 946-80-5 Molecular Formula: C6H5CH2OC6H5 Formula Weight: 184.24 Flash Point: 110°(230°F) Melting Point: 37-40° Boiling Point: 286-287° Beilstein Reference: 2045713 MDL No.: MFCD00020660

Formula: C₁₃H₁₂O
Boiling Pt: 286…287 °C
Melting Pt: 37…40 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00020660
CAS Number: 946-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR