5-Chloro-1,3-benzodioxole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Chlorobenzo[d][1,3]dioxole

B22534-09 B22534-18
CAAAB22534-09EA 71.26 CAD
CAAAB22534-09 CAAAB22534-18 CAAAB22534-30
5-Chloro-1,3-benzodioxole 98%
5-Chloro-1,3-benzodioxole

Purity: 98% CAS No.: 7228-38-8 Molecular Formula: C7H5ClO2 Formula Weight: 156.57 Flash Point: 93°(201°F) Refractive Index: 1.5560 Boiling Point: 185-187° MDL No.: MFCD00010842

Formula: C₇H₅ClO₂
MW: 156.57 g/mol
Boiling Pt: 185…187 °C
Density: 1.340
Flash Pt: 93 °C(201 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010842
CAS Number: 7228-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR