5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone 98%

Supplier: Thermo Scientific Chemicals
Warning

B22521-09 B22521-18
CAAAB22521-09EA 199.08 CAD
CAAAB22521-09 CAAAB22521-18
5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone 98%
5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone

Purity: 98% CAS No.: 53786-28-0 Molecular Formula: C12H14ClN3O Formula Weight: 251.71 Melting Point: 227-230° MDL No.: MFCD02684518

Formula: C₁₂H₁₄ClN₃O
Melting Pt: 227…230 °C
Storage Temperature: Ambient
MDL Number: MFCD02684518
CAS Number: 53786-28-0
EINECS: 258-771-6
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR